Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGGGTCCTAGAAATTTGCTTG[A/C]CAGGTGAGCCCTCCGTTGGGCCGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHD1 - pleckstrin homology and coiled-coil domain containing D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161498.1 | Intron | NP_001154970.1 | ||||
XM_011536762.2 | Intron | XP_011535064.1 | ||||
XM_011536763.2 | Intron | XP_011535065.1 | ||||
XM_017021290.1 | Intron | XP_016876779.1 |