Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACGGCAGAGGGAAGGGAGGAAA[A/C]AGTGCAGGGAAGGGAGAGGGCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601672 MIM: 603894 | ||||||||||||||||||||
Literature Links: |
DPF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPF3 - double PHD fingers 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280542.1 | Intron | NP_001267471.1 | ||||
NM_001280543.1 | Intron | NP_001267472.1 | ||||
NM_001280544.1 | Intron | NP_001267473.1 | ||||
NM_012074.4 | Intron | NP_036206.3 | ||||
XM_017021670.1 | Intron | XP_016877159.1 | ||||
XM_017021671.1 | Intron | XP_016877160.1 | ||||
XM_017021672.1 | Intron | XP_016877161.1 | ||||
XM_017021673.1 | Intron | XP_016877162.1 | ||||
XM_017021674.1 | Intron | XP_016877163.1 | ||||
XM_017021675.1 | Intron | XP_016877164.1 |