Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTAGAGTGCTATCAGAAGAGT[C/T]TGTTTCATGGAAGGAGAAACACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH6A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH6A1 - aldehyde dehydrogenase 6 family member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIN52 - lin-52 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024674.2 | Intron | NP_001019845.1 | ||||
XM_011537320.2 | Intron | XP_011535622.1 | ||||
XM_011537321.2 | Intron | XP_011535623.1 | ||||
XM_011537322.2 | Intron | XP_011535624.2 | ||||
XM_017021763.1 | Intron | XP_016877252.1 | ||||
XM_017021764.1 | Intron | XP_016877253.1 |