Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGACTCCTTCACTTGCAGCAAA[A/G]TATTGAAATTCCGGAGATCAGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600112 | ||||||||||||||||||||
Literature Links: |
DYNC1H1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYNC1H1 - dynein cytoplasmic 1 heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001376.4 | 760 | Missense Mutation | AAT,AGT | N,S 199 | NP_001367.2 |