Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCCTCCTGTCCCAAGAGGTATT[C/T]ATGGAGCGCGCGGCCCTGCCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612498 MIM: 605567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADSSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADSSL1 - adenylosuccinate synthase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIVA1 - SIVA1 apoptosis inducing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006427.3 | Intron | NP_006418.2 | ||||
NM_021709.2 | Intron | NP_068355.1 | ||||
XM_011536360.2 | Intron | XP_011534662.1 |