Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTTAATTTTGAGCTGTGAAGA[T/C]CTCCTCCGTAGATGGGTCTTCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137192 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRB3 - gamma-aminobutyric acid type A receptor beta3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000814.5 | 1503 | Silent Mutation | AGA,AGG | R,R 431 | NP_000805.1 | |
NM_001191320.1 | 1503 | Silent Mutation | AGA,AGG | R,R 346 | NP_001178249.1 | |
NM_001191321.2 | 1503 | Silent Mutation | AGA,AGG | R,R 360 | NP_001178250.1 | |
NM_001278631.1 | 1503 | Silent Mutation | AGA,AGG | R,R 346 | NP_001265560.1 | |
NM_021912.4 | 1503 | Silent Mutation | AGA,AGG | R,R 431 | NP_068712.1 | |
XM_011521428.2 | 1503 | Silent Mutation | AGA,AGG | R,R 372 | XP_011519730.1 |