Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGCATCCAGTCTTATGACTCT[A/G]CTGTTGTCTTTGCTGTTCTCTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616670 MIM: 609931 MIM: 609930 MIM: 601734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESYT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESYT1 - extended synaptotagmin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107987175 - uncharacterized LOC107987175 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017020294.1 | Intron | XP_016875783.1 |
MYL6 - myosin light chain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL6B - myosin light chain 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199629.1 | Intron | NP_001186558.1 | ||||
NM_002475.4 | Intron | NP_002466.1 |
SMARCC2 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |