Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTCTATGATAGCAAGTGACAC[A/G]TCCTCTTGATCCACTTCAAGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM19A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM19A2 - family with sequence similarity 19 member A2, C-C motif chemokine like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178539.4 | Intron | NP_848634.1 | ||||
XM_006719370.2 | Intron | XP_006719433.1 | ||||
XM_011538272.1 | Intron | XP_011536574.1 | ||||
XM_011538274.1 | Intron | XP_011536576.1 |