Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATTTTCTCTCAATTTCTCATTT[C/T]CAGGAATGTGTATTTATTTATTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP2B1 - ATPase plasma membrane Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001323.1 | Intron | NP_001001323.1 | ||||
NM_001682.2 | Intron | NP_001673.2 | ||||
XM_011538407.2 | Intron | XP_011536709.1 | ||||
XM_017019357.1 | Intron | XP_016874846.1 | ||||
XM_017019358.1 | Intron | XP_016874847.1 | ||||
XM_017019359.1 | Intron | XP_016874848.1 | ||||
XM_017019360.1 | Intron | XP_016874849.1 |