Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATATGGCCTATTTTAGAAAGCA[G/T]AGTCAGGGAATGTGTCTCTCATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH1L2 - aldehyde dehydrogenase 1 family member L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034173.3 | 6190 | UTR 3 | NP_001029345.2 | |||
XM_011537986.2 | 6190 | UTR 3 | XP_011536288.1 | |||
XM_011537988.2 | 6190 | Intron | XP_011536290.1 | |||
XM_011537989.2 | 6190 | Intron | XP_011536291.1 | |||
XM_017018889.1 | 6190 | UTR 3 | XP_016874378.1 | |||
XM_017018890.1 | 6190 | Intron | XP_016874379.1 |