Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAAGGGCGGAGAAGAGGGACA[C/T]GTGCCAGACAACAGCTGCAGAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TCTEX1D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCTEX1D2 - Tctex1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM4SF19 - transmembrane 4 L six family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204897.1 | 678 | Silent Mutation | ACA,ACG | T,T 172 | NP_001191826.1 | |
NM_001204898.1 | 678 | Missense Mutation | ATG,GTG | M,V 148 | NP_001191827.1 | |
NM_138461.3 | 678 | Missense Mutation | ATG,GTG | M,V 174 | NP_612470.2 |
TM4SF19-AS1 - TM4SF19 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM4SF19-TCTEX1D2 - TM4SF19-TCTEX1D2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |