Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGACACACAGACTGTTCTGTG[A/G]ATGCTGAGGGTTTGCTGGGCTTTAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615322 | |||||||||||||||||||||||
Literature Links: |
LINC01063 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LINC01063 - long intergenic non-protein coding RNA 1063 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRROS - negative regulator of reactive oxygen species | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198565.2 | Intron | NP_940967.1 |