Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTGTATTATAAGAAGAATAAA[A/G]TAGCTGATGTTAATAATCTGTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCU - mitochondrial calcium uniporter | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270679.1 | Intron | NP_001257608.1 | ||||
NM_001270680.1 | Intron | NP_001257609.1 | ||||
NM_138357.2 | Intron | NP_612366.1 | ||||
XM_017016882.1 | Intron | XP_016872371.1 |
MIR4676 - microRNA 4676 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |