Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAAAAGCCCAGCAAACAGGAGC[G/A]TTTTACCTCTTTCATAGTCTGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609233 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT13 - nudix hydrolase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283014.1 | 418 | Silent Mutation | GCA,GCG | A,A 48 | NP_001269943.1 | |
NM_001283015.1 | 418 | Silent Mutation | GCA,GCG | A,A 48 | NP_001269944.1 | |
NM_001283016.1 | 418 | UTR 5 | NP_001269945.1 | |||
NM_001283017.1 | 418 | UTR 5 | NP_001269946.1 | |||
NM_001283019.1 | 418 | UTR 5 | NP_001269948.1 | |||
NM_015901.5 | 418 | Silent Mutation | GCA,GCG | A,A 48 | NP_056985.3 | |
XM_017016033.1 | 418 | Silent Mutation | GCA,GCG | A,A 48 | XP_016871522.1 |