Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGGTGAAGGCATCTACTGGCAC[A/G]TCTTGTAGAGGGGGGATCCCTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610454 MIM: 612971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZTS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LZTS2 - leucine zipper tumor suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD7 - PDZ domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195263.1 | 2379 | Silent Mutation | GAC,GAT | D,D 719 | NP_001182192.1 | |
NM_024895.4 | 2379 | Intron | NP_079171.1 | |||
XM_005270165.2 | 2379 | Intron | XP_005270222.1 | |||
XM_011540177.2 | 2379 | Silent Mutation | GAC,GAT | D,D 719 | XP_011538479.1 | |
XM_011540178.2 | 2379 | Silent Mutation | GAC,GAT | D,D 718 | XP_011538480.1 | |
XM_011540179.2 | 2379 | Intron | XP_011538481.1 | |||
XM_011540181.2 | 2379 | Intron | XP_011538483.1 | |||
XM_011540182.2 | 2379 | Intron | XP_011538484.1 | |||
XM_011540183.2 | 2379 | Intron | XP_011538485.1 | |||
XM_017016667.1 | 2379 | Intron | XP_016872156.1 | |||
XM_017016668.1 | 2379 | Intron | XP_016872157.1 |