Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGCAGCCTCAGCGCTCGCGGCC[A/G]GGGGATCCTGCAGGCGGCTGCGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNNM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNNM2 - cyclin and CBS domain divalent metal cation transport mediator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017649.4 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | NP_060119.3 | |
NM_199076.2 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | NP_951058.1 | |
NM_199077.2 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | NP_951059.1 | |
XM_005269933.4 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | XP_005269990.1 | |
XM_006717908.2 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | XP_006717971.1 | |
XM_011539911.2 | 301 | Missense Mutation | CAG,CGG | Q,R 38 | XP_011538213.1 |
LOC107984265 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017017027.1 | 301 | Intron | XP_016872516.1 |