Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAAGTTGTGCCTCTTAAACAGA[C/T]TTCTTAGTCTCTTTCAACTTTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VTI1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VTI1A - vesicle transport through interaction with t-SNAREs 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318203.1 | 260 | Intron | NP_001305132.1 | |||
NM_001318205.1 | 260 | Intron | NP_001305134.1 | |||
NM_145206.3 | 260 | Intron | NP_660207.2 | |||
XM_005269544.4 | 260 | Intron | XP_005269601.1 | |||
XM_005269545.4 | 260 | Intron | XP_005269602.1 | |||
XM_005269546.3 | 260 | Intron | XP_005269603.1 | |||
XM_005269547.4 | 260 | Intron | XP_005269604.1 | |||
XM_006717637.2 | 260 | Intron | XP_006717700.1 | |||
XM_011539328.2 | 260 | Intron | XP_011537630.1 | |||
XM_011539330.2 | 260 | Intron | XP_011537632.1 | |||
XM_011539331.2 | 260 | Intron | XP_011537633.1 | |||
XM_011539333.2 | 260 | Intron | XP_011537635.1 | |||
XM_017015745.1 | 260 | Intron | XP_016871234.1 | |||
XM_017015746.1 | 260 | Intron | XP_016871235.1 | |||
XM_017015747.1 | 260 | Intron | XP_016871236.1 | |||
XM_017015748.1 | 260 | Intron | XP_016871237.1 |
ZDHHC6 - zinc finger DHHC-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303134.1 | 260 | Intron | NP_001290063.1 | |||
NM_022494.2 | 260 | Intron | NP_071939.1 | |||
XM_005270051.4 | 260 | UTR 5 | XP_005270108.1 | |||
XM_006717948.3 | 260 | Intron | XP_006718011.1 | |||
XM_006717949.3 | 260 | Intron | XP_006718012.1 | |||
XM_011540091.2 | 260 | Intron | XP_011538393.1 | |||
XM_011540092.2 | 260 | Intron | XP_011538394.1 | |||
XM_011540093.1 | 260 | Intron | XP_011538395.1 | |||
XM_011540094.2 | 260 | Intron | XP_011538396.1 | |||
XM_011540095.2 | 260 | Intron | XP_011538397.1 | |||
XM_017016561.1 | 260 | Intron | XP_016872050.1 | |||
XM_017016562.1 | 260 | Intron | XP_016872051.1 | |||
XM_017016563.1 | 260 | Intron | XP_016872052.1 | |||
XM_017016564.1 | 260 | Intron | XP_016872053.1 | |||
XM_017016565.1 | 260 | Intron | XP_016872054.1 | |||
XM_017016566.1 | 260 | Intron | XP_016872055.1 | |||
XM_017016567.1 | 260 | Intron | XP_016872056.1 | |||
XM_017016568.1 | 260 | Intron | XP_016872057.1 | |||
XM_017016569.1 | 260 | Intron | XP_016872058.1 |