Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAATGGTCTTAACTATCTCTTTT[A/G]TGGTAGATATGTAACTACTTGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF684 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF684 - zinc finger protein 684 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152373.3 | Intron | NP_689586.3 | ||||
XM_011540671.2 | Intron | XP_011538973.1 | ||||
XM_011540672.2 | Intron | XP_011538974.1 | ||||
XM_011540673.2 | Intron | XP_011538975.1 | ||||
XM_017000290.1 | Intron | XP_016855779.1 | ||||
XM_017000291.1 | Intron | XP_016855780.1 | ||||
XM_017000292.1 | Intron | XP_016855781.1 |