Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGGGGAACAAGAGATGAGCTC[A/G]TAAGGGCACAGTCTGGGGCAGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 186357 | ||||||||||||||||||||
Literature Links: |
SDC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDC3 - syndecan 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014654.3 | 1586 | UTR 3 | NP_055469.3 | |||
XM_011542463.1 | 1586 | UTR 3 | XP_011540765.1 | |||
XM_011542464.2 | 1586 | UTR 3 | XP_011540766.1 | |||
XM_011542466.1 | 1586 | UTR 3 | XP_011540768.1 |