Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGCTCCCCCCTGCCGGAAGCC[G/A]GAAGCGATCTCATCGAAGGTCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138140 MIM: 143090 | ||||||||||||||||||||
Literature Links: |
SLC2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC2A1 - solute carrier family 2 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006516.2 | 1920 | Silent Mutation | TCC,TCT | S,S 465 | NP_006507.2 |