Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGGATCCTAAGTCATCCTCGG[C/T]CTCAGCCTCTGCTGTGACAAGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1522 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1522 - KIAA1522 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198972.1 | Intron | NP_001185901.1 | ||||
NM_001198973.1 | Intron | NP_001185902.1 | ||||
NM_020888.2 | Intron | NP_065939.2 | ||||
XM_006710791.3 | Intron | XP_006710854.1 | ||||
XM_017001916.1 | Intron | XP_016857405.1 | ||||
XM_017001917.1 | Intron | XP_016857406.1 |