Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAACAGACATGATGAATCTGGC[A/G]CTGGGAGTTCCTGGAAATTCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608353 MIM: 616755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AZIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM62 - tripartite motif containing 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018207.2 | 3126 | UTR 3 | NP_060677.2 | |||
XM_011541705.2 | 3126 | UTR 3 | XP_011540007.1 | |||
XM_017001629.1 | 3126 | UTR 3 | XP_016857118.1 | |||
XM_017001630.1 | 3126 | UTR 3 | XP_016857119.1 | |||
XM_017001631.1 | 3126 | Intron | XP_016857120.1 |