Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAATTATAAAGTTAACCCTTAA[A/G]TGGCCAATGACACATAATAGATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613121 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEXN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEXN - nexilin F-actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172309.1 | Intron | NP_001165780.1 | ||||
NM_144573.3 | Intron | NP_653174.3 | ||||
XM_005271322.3 | Intron | XP_005271379.1 | ||||
XM_005271323.3 | Intron | XP_005271380.1 | ||||
XM_005271324.4 | Intron | XP_005271381.1 | ||||
XM_005271325.3 | Intron | XP_005271382.1 | ||||
XM_005271326.3 | Intron | XP_005271383.1 | ||||
XM_005271327.3 | Intron | XP_005271384.1 |