Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTACTTCCATGCATCTTCCAC[A/G]TATTGTGGTATTTTCCATGTATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC8B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC8B - leucine rich repeat containing 8 family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134476.1 | Intron | NP_001127948.1 | ||||
NM_015350.2 | Intron | NP_056165.1 | ||||
XM_005270701.4 | Intron | XP_005270758.1 | ||||
XM_005270702.4 | Intron | XP_005270759.1 | ||||
XM_005270703.4 | Intron | XP_005270760.1 | ||||
XM_011541143.2 | Intron | XP_011539445.1 | ||||
XM_011541144.2 | Intron | XP_011539446.1 | ||||
XM_011541145.2 | Intron | XP_011539447.1 | ||||
XM_011541146.2 | Intron | XP_011539448.1 | ||||
XM_011541148.2 | Intron | XP_011539450.1 | ||||
XM_017000883.1 | Intron | XP_016856372.1 | ||||
XM_017000884.1 | Intron | XP_016856373.1 | ||||
XM_017000885.1 | Intron | XP_016856374.1 | ||||
XM_017000886.1 | Intron | XP_016856375.1 |