Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTGTACAGACATGGCGGCGG[C/G]TTTTCGGAAGGCGGCTAAGTCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602790 MIM: 609440 | ||||||||||||||||||||
Literature Links: |
FHL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FHL3 - four and a half LIM domains 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP11 - UTP11, small subunit processome component homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016037.3 | 98 | Missense Mutation | GCT,GGT | A,G 4 | NP_057121.2 | |
XM_011541555.2 | 98 | UTR 5 | XP_011539857.1 |