Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTTATGGACATAAAGACTCTA[C/T]GCAGAAACCTCCTCAACTTTAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609844 MIM: 159350 | ||||||||||||||||||||
Literature Links: |
DCP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCP2 - decapping mRNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCC - mutated in colorectal cancers | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085377.1 | 4526 | UTR 3 | NP_001078846.1 | |||
NM_002387.2 | 4526 | UTR 3 | NP_002378.1 | |||
XM_005271991.2 | 4526 | UTR 3 | XP_005272048.1 | |||
XM_017009473.1 | 4526 | UTR 3 | XP_016864962.1 | |||
XM_017009474.1 | 4526 | UTR 3 | XP_016864963.1 |