Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATCTTGAGAGAGAGGGAGGCAG[A/G]CAGGACCTGTGGCTGTGATCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605604 MIM: 603790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAIP2 - poly(A) binding protein interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC23A1 - solute carrier family 23 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005847.4 | Intron | NP_005838.3 | ||||
NM_152685.3 | Intron | NP_689898.2 | ||||
XM_005272148.3 | Intron | XP_005272205.3 | ||||
XM_005272149.3 | Intron | XP_005272206.3 | ||||
XM_006714741.2 | Intron | XP_006714804.2 | ||||
XM_011543765.2 | Intron | XP_011542067.1 | ||||
XM_011543766.1 | Intron | XP_011542068.1 | ||||
XM_011543767.1 | Intron | XP_011542069.1 | ||||
XM_011543768.1 | Intron | XP_011542070.1 | ||||
XM_011543769.1 | Intron | XP_011542071.1 |