Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTTTGCCGCAAGGCTCTGCACC[A/G]TCAAAGAGAGTAGGGCACTGAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOMER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOMER2 - homer scaffolding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004839.3 | 5787 | Intron | NP_004830.2 | |||
NM_199330.2 | 5787 | Intron | NP_955362.1 | |||
XM_005272448.4 | 5787 | UTR 3 | XP_005272505.1 | |||
XM_005272449.3 | 5787 | UTR 3 | XP_005272506.1 | |||
XM_006720775.3 | 5787 | UTR 3 | XP_006720838.1 | |||
XM_006720776.3 | 5787 | UTR 3 | XP_006720839.1 | |||
XM_011522231.2 | 5787 | UTR 3 | XP_011520533.1 | |||
XM_011522232.2 | 5787 | Intron | XP_011520534.1 | |||
XM_011522233.2 | 5787 | UTR 3 | XP_011520535.1 | |||
XM_011522234.2 | 5787 | Intron | XP_011520536.1 |