Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGGCTGTCCTAAAACCACAAG[C/G]TGGGTTAGGTAAGTGTTTTGTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKAB2 - protein kinase AMP-activated non-catalytic subunit beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005399.4 | 5075 | UTR 3 | NP_005390.1 | |||
XM_011509729.1 | 5075 | UTR 3 | XP_011508031.1 | |||
XM_017001697.1 | 5075 | UTR 3 | XP_016857186.1 |