Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGATTAAGCTTCAAATACAAAT[A/G]TGGAGGGAAATCTATACATATCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIN9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIN9 - lin-9 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270409.1 | Intron | NP_001257338.1 | ||||
NM_001270410.1 | Intron | NP_001257339.1 | ||||
NM_173083.3 | Intron | NP_775106.2 | ||||
XM_005273102.3 | Intron | XP_005273159.1 | ||||
XM_006711766.2 | Intron | XP_006711829.2 | ||||
XM_006711767.3 | Intron | XP_006711830.1 | ||||
XM_011544173.2 | Intron | XP_011542475.1 | ||||
XM_017001084.1 | Intron | XP_016856573.1 |