Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCCTGTTTGAGGATGCAGACT[C/T]CCCAGCCAGCAATTCCTCCCTGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606401 | ||||||||||||||||||||
Literature Links: |
CAPN9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN9 - calpain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319676.1 | 240 | Missense Mutation | TCC,TTC | S,F 48 | NP_001306605.1 | |
NM_006615.2 | 240 | Missense Mutation | TCC,TTC | S,F 48 | NP_006606.1 | |
NM_016452.2 | 240 | Missense Mutation | TCC,TTC | S,F 48 | NP_057536.1 | |
XM_011544017.1 | 240 | Missense Mutation | TCC,TTC | S,F 48 | XP_011542319.1 | |
XM_011544018.1 | 240 | Missense Mutation | TCC,TTC | S,F 48 | XP_011542320.1 | |
XM_011544019.2 | 240 | Missense Mutation | TCC,TTC | S,F 48 | XP_011542321.1 | |
XM_011544020.1 | 240 | Intron | XP_011542322.1 | |||
XM_017000098.1 | 240 | Missense Mutation | TCC,TTC | S,F 48 | XP_016855587.1 | |
XM_017000099.1 | 240 | Intron | XP_016855588.1 |