Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTACTGGCGTCCCCGGCCTCTG[A/C]CTGGGTGACTCGGGAGGGCAAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK1 - potassium two pore domain channel subfamily K member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002245.3 | Intron | NP_002236.1 | ||||
XM_011544184.2 | Intron | XP_011542486.1 |
MIR4427 - microRNA 4427 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |