Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAATAAAATATTCAGTATTTTAT[G/T]AACAATTCAGAAGTCTGTGTAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID4B - AT-rich interaction domain 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206794.1 | Intron | NP_001193723.1 | ||||
NM_016374.5 | Intron | NP_057458.4 | ||||
NM_031371.3 | Intron | NP_112739.2 | ||||
XM_006711781.2 | Intron | XP_006711844.1 | ||||
XM_011544212.2 | Intron | XP_011542514.1 | ||||
XM_017001468.1 | Intron | XP_016856957.1 | ||||
XM_017001469.1 | Intron | XP_016856958.1 | ||||
XM_017001470.1 | Intron | XP_016856959.1 | ||||
XM_017001471.1 | Intron | XP_016856960.1 | ||||
XM_017001472.1 | Intron | XP_016856961.1 |