Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGCTGTCATTGGTGGTGGCTT[G/T]GTAAGAATTTTCAGATGGATTACTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603538 | ||||||||||||||||||||
Literature Links: |
KMO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KMO - kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003679.4 | 475 | Missense Mutation | TTG,TTT | L,F 18 | NP_003670.2 | |
XM_005273337.1 | 475 | Missense Mutation | TTG,TTT | L,F 58 | XP_005273394.1 | |
XM_005273338.3 | 475 | UTR 5 | XP_005273395.1 | |||
XM_017002649.1 | 475 | UTR 5 | XP_016858138.1 | |||
XM_017002650.1 | 475 | Intron | XP_016858139.1 |