Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGATTTTATGTGGATGATTTGC[A/C]AGAAAATTTTTCTTGCCATAGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613272 MIM: 608400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD3 - potassium channel tetramerization domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USH2A - usherin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007123.5 | 17184 | Intron | NP_009054.5 | |||
NM_206933.2 | 17184 | UTR 3 | NP_996816.2 |