Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCTGACTAAGTGTCCATGTTC[A/G]TGTGACTTGCAGGCAAGCCAGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601671 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPF2 - double PHD fingers 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006268.4 | Intron | NP_006259.1 | ||||
XM_005274149.2 | Intron | XP_005274206.1 | ||||
XM_017018101.1 | Intron | XP_016873590.1 | ||||
XM_017018102.1 | Intron | XP_016873591.1 |