Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCTGAGCCGGAAGGGGCCATG[A/G]GCAGAAGGAGGGGAGCGCCTGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT11 - Wnt family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004626.2 | Intron | NP_004617.2 | ||||
XM_005274231.1 | Intron | XP_005274288.1 | ||||
XM_011545238.1 | Intron | XP_011543540.1 | ||||
XM_011545239.2 | Intron | XP_011543541.1 | ||||
XM_011545240.2 | Intron | XP_011543542.1 | ||||
XM_011545241.2 | Intron | XP_011543543.1 | ||||
XM_017018241.1 | Intron | XP_016873730.1 | ||||
XM_017018242.1 | Intron | XP_016873731.1 | ||||
XM_017018243.1 | Intron | XP_016873732.1 |