Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTAGAACCAACATATTCAGAA[A/G]TGGGCATGGACACTAGGGCCACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYATL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLYATL1 - glycine-N-acyltransferase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220494.2 | Intron | NP_001207423.1 | ||||
NM_001220496.2 | Intron | NP_001207425.1 | ||||
NM_080661.4 | Intron | NP_542392.2 | ||||
XM_011545354.2 | Intron | XP_011543656.1 | ||||
XM_011545355.2 | Intron | XP_011543657.1 | ||||
XM_011545356.2 | Intron | XP_011543658.1 | ||||
XM_011545357.2 | Intron | XP_011543659.1 | ||||
XM_011545358.2 | Intron | XP_011543660.1 | ||||
XM_011545359.2 | Intron | XP_011543661.1 |
LOC283194 - uncharacterized LOC283194 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |