Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTATAAACTTCCTTCTTAGTAC[A/G]GCTTTTGTCGCATCTCATAATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPSF7 - cleavage and polyadenylation specific factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136040.2 | Intron | NP_001129512.1 | ||||
NM_001142565.1 | Intron | NP_001136037.1 | ||||
NM_024811.3 | Intron | NP_079087.3 | ||||
XM_005274298.4 | Intron | XP_005274355.1 | ||||
XM_005274299.4 | Intron | XP_005274356.1 | ||||
XM_005274303.4 | Intron | XP_005274360.1 | ||||
XM_011545257.2 | Intron | XP_011543559.1 | ||||
XM_011545258.2 | Intron | XP_011543560.1 | ||||
XM_011545259.2 | Intron | XP_011543561.1 | ||||
XM_011545260.2 | Intron | XP_011543562.1 | ||||
XM_011545261.2 | Intron | XP_011543563.1 | ||||
XM_011545262.2 | Intron | XP_011543564.1 | ||||
XM_011545263.2 | Intron | XP_011543565.1 | ||||
XM_017018345.1 | Intron | XP_016873834.1 |
SDHAF2 - succinate dehydrogenase complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017841.2 | Intron | NP_060311.1 |