Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGCAAAGAGCACAGGAAAGAT[G/A]ATGCGGGAAATTTTGTCAACCTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 305660 | ||||||||||||||||||||
Literature Links: |
GABRA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABRA3 - gamma-aminobutyric acid type A receptor alpha3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000808.3 | 1411 | Silent Mutation | ATC,ATT | I,I 461 | NP_000799.1 | |
XM_006724811.2 | 1411 | UTR 3 | XP_006724874.1 |