Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCTGGCCAGAGTAACTACGGT[C/T]CACTAAAACTATTCTGGATTAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMPK2 - cytidine/uridine monophosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256477.1 | 1466 | Intron | NP_001243406.1 | |||
NM_001256478.1 | 1466 | Intron | NP_001243407.1 | |||
NM_207315.3 | 1466 | Missense Mutation | GAA,GGA | E,G 448 | NP_997198.2 |
NRIR - negative regulator of interferon response (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |