Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGGGTTTGTTTGTTTCCCAAGA[G/T]AAAATATTTTAACTTTCAGAAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF144A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF144A - ring finger protein 144A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014746.4 | Intron | NP_055561.2 | ||||
XM_005246200.3 | Intron | XP_005246257.1 | ||||
XM_005246202.4 | Intron | XP_005246259.1 | ||||
XM_017005396.1 | Intron | XP_016860885.1 | ||||
XM_017005397.1 | Intron | XP_016860886.1 | ||||
XM_017005398.1 | Intron | XP_016860887.1 | ||||
XM_017005399.1 | Intron | XP_016860888.1 | ||||
XM_017005400.1 | Intron | XP_016860889.1 | ||||
XM_017005401.1 | Intron | XP_016860890.1 | ||||
XM_017005402.1 | Intron | XP_016860891.1 | ||||
XM_017005403.1 | Intron | XP_016860892.1 | ||||
XM_017005404.1 | Intron | XP_016860893.1 |
RNF144A-AS1 - RNF144A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |