Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAAAGATTCTAAGTCTAGCTGA[C/T]GAGAGCCTGTAGTGAGTGATGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606029 MIM: 607153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPSF3 - cleavage and polyadenylation specific factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321833.1 | Intron | NP_001308762.1 | ||||
NM_001321834.1 | Intron | NP_001308763.1 | ||||
NM_001321835.1 | Intron | NP_001308764.1 | ||||
NM_001321836.1 | Intron | NP_001308765.1 | ||||
NM_016207.3 | Intron | NP_057291.1 |
ITGB1BP1 - integrin subunit beta 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |