Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTGGATGTGGGACTCTGAGACT[C/T]CTCTGCTTCTGTGACCTTTGTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610318 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COBLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COBLL1 - cordon-bleu WH2 repeat protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278458.1 | 7546 | UTR 3 | NP_001265387.1 | |||
NM_001278460.1 | 7546 | UTR 3 | NP_001265389.1 | |||
NM_001278461.1 | 7546 | UTR 3 | NP_001265390.1 | |||
NM_014900.4 | 7546 | UTR 3 | NP_055715.3 |
SNORA70F - small nucleolar RNA, H/ACA box 70F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |