Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAATGCTCATTCCATCCACAGA[G/T]TTGCAGAATTAGAGGTTTTATTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 161650 MIM: 608952 | ||||||||||||||||||||
Literature Links: |
NEB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIF1 - replication timing regulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177663.1 | 26097 | Intron | NP_001171134.1 | |||
NM_001177664.1 | 26097 | Intron | NP_001171135.1 | |||
NM_001177665.1 | 26097 | Intron | NP_001171136.1 | |||
NM_018151.4 | 26097 | Intron | NP_060621.3 | |||
XM_005246665.3 | 26097 | Intron | XP_005246722.1 | |||
XM_011511395.2 | 26097 | Intron | XP_011509697.1 | |||
XM_017004422.1 | 26097 | Intron | XP_016859911.1 | |||
XM_017004423.1 | 26097 | Intron | XP_016859912.1 | |||
XM_017004424.1 | 26097 | Intron | XP_016859913.1 | |||
XM_017004425.1 | 26097 | Intron | XP_016859914.1 |