Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTCTGGATGGCAGGTTAGCTGT[A/G]TGCTCTTTGGGAGAACTGAGCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616285 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMNL2 - formin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052905.3 | Intron | NP_443137.2 | ||||
XM_005246263.2 | Intron | XP_005246320.1 | ||||
XM_005246265.3 | Intron | XP_005246322.1 | ||||
XM_011510530.1 | Intron | XP_011508832.1 | ||||
XM_011510531.1 | Intron | XP_011508833.1 | ||||
XM_011510532.2 | Intron | XP_011508834.1 | ||||
XM_011510533.2 | Intron | XP_011508835.1 | ||||
XM_011510534.1 | Intron | XP_011508836.1 | ||||
XM_011510535.2 | Intron | XP_011508837.1 | ||||
XM_011510536.2 | Intron | XP_011508838.1 |