Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAGATCCAGACGGTTAAAACA[A/T]GTATGCGCTCTGAAAACACAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HECW2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HECW2 - HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304840.1 | 4837 | Silent Mutation | ACA,ACT | T,T 1183 | NP_001291769.1 | |
NM_020760.2 | 4837 | Silent Mutation | ACA,ACT | T,T 1539 | NP_065811.1 | |
XM_006712646.3 | 4837 | Silent Mutation | ACA,ACT | T,T 1546 | XP_006712709.1 | |
XM_006712648.3 | 4837 | Silent Mutation | ACA,ACT | T,T 1415 | XP_006712711.1 | |
XM_017004563.1 | 4837 | Silent Mutation | ACA,ACT | T,T 1539 | XP_016860052.1 |