Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCTGAATTTAAAATCCCATCAA[A/C]ACTTTTTTGCTCTTAAGCATAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604155 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LANCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LANCL1 - LanC like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136574.1 | 3545 | UTR 3 | NP_001130046.1 | |||
NM_001136575.1 | 3545 | UTR 3 | NP_001130047.1 | |||
NM_006055.2 | 3545 | UTR 3 | NP_006046.1 | |||
XM_005246243.2 | 3545 | UTR 3 | XP_005246300.1 |
LANCL1-AS1 - LANCL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |