Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTATGAACAAATTACAGCATGGT[A/G]GAACAGAAGAAGGTATATGAGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSL3 - acyl-CoA synthetase long-chain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004457.3 | Intron | NP_004448.2 | ||||
NM_203372.1 | Intron | NP_976251.1 | ||||
XM_017003584.1 | Intron | XP_016859073.1 |