Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAAAAGAAAACAGCTTGTCCAT[A/G]TAGAAAGATTAGAGGATCTAAATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610072 MIM: 615129 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERMN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERMN - ermin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009959.2 | 2696 | UTR 3 | NP_001009959.1 | |||
NM_001304344.1 | 2696 | UTR 3 | NP_001291273.1 | |||
NM_001304345.1 | 2696 | UTR 3 | NP_001291274.1 | |||
NM_001304346.1 | 2696 | UTR 3 | NP_001291275.1 | |||
NM_020711.2 | 2696 | UTR 3 | NP_065762.1 |
GALNT5 - polypeptide N-acetylgalactosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |